The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166621.1 | NP_001160093.1 | 2 - 3 | 156 | 63 |
NM_021210.4 | NP_067033.1 | 1 - 2 | 216 | 63 | |
GenBank mRNA | AK314038.1 | - | 1 - 2 | 222 | 63 |
BC032717.1 | - | 1 - 2 | 218 | 63 | |
BI457771.1 | - | 1 - 2 | 218 | 63 | |
BI600633.1 | - | 2 - 3 | 161 | 63 | |
BM762006.1 | - | 2 - 3 | 130 | 63 |