Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DUS4L
Entrez Gene ID
11062
Gene Name
dihydrouridine synthase 4 like
Gene Aliases
DUS4, PP35
Chromosome Location
Chr.7: 107563957 - 107578523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270419.1 NP_001257348.1 1 - 2 103 122
NR_073005.1 1 - 2 103 122
GenBank mRNA AL529385.3 - 1 - 2 80 122

View Full Product Details