Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCF2
Entrez Gene ID
4168
Gene Name
MCF.2 cell line derived transforming sequence
Gene Aliases
ARHGEF21, DBL
Chromosome Location
Chr.X: 139581768 - 139708279 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099855.1 NP_001093325.1 14 - 15 1735 126
NM_001171876.1 NP_001165347.1 15 - 16 1783 174
NM_001171877.1 NP_001165348.1 10 - 11 1600 126
NM_001171878.1 NP_001165349.1 11 - 12 1717 126
NM_001171879.1 NP_001165350.1 12 - 13 1765 174
NM_005369.4 NP_005360.3 11 - 12 1717 126
XM_005262413.4 XP_005262470.1 16 - 17 1931 174
XM_011531339.2 XP_011529641.1 16 - 17 1990 174
XM_017029529.1 XP_016885018.1 15 - 16 1752 174
XM_017029530.1 XP_016885019.1 15 - 16 1752 174
XM_017029531.1 XP_016885020.1 15 - 16 1685 174
XM_017029532.1 XP_016885021.1 12 - 13 1842 174
GenBank mRNA AB085901.1 - 11 - 12 1394 126
AB085902.1 - 12 - 13 1442 174
AK296488.1 - 10 - 11 1600 126
AK296529.1 - 14 - 15 1835 126
AK302957.1 - 16 - 17 1731 174
AK313900.1 - 11 - 12 1577 126
AK316416.1 - 15 - 16 1771 174
AL117234.1 - 13 - 14 1618 126
X12556.1 - 11 - 12 1568 126
X13230.1 - - 322 126

View Full Product Details