The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000246.3 | NP_000237.2 | 6 - 7 | 619 | 134 |
NM_001286402.1 | NP_001273331.1 | 6 - 7 | 622 | 134 | |
NR_104444.1 | 6 - 7 | 619 | 134 | ||
XM_006720880.3 | XP_006720943.2 | 6 - 7 | 800 | 134 | |
XM_011522484.2 | XP_011520786.1 | 6 - 7 | 800 | 134 | |
XM_011522485.2 | XP_011520787.1 | 6 - 7 | 800 | 134 | |
XM_011522486.2 | XP_011520788.1 | 6 - 7 | 802 | 134 | |
XM_011522488.2 | XP_011520790.1 | 6 - 7 | 2031 | 134 | |
XM_011522490.2 | XP_011520792.1 | 6 - 7 | 2029 | 134 | |
XM_011522491.2 | XP_011520793.1 | 6 - 7 | 802 | 134 | |
XM_011522492.2 | XP_011520794.1 | 6 - 7 | 748 | 134 | |
XM_011522493.2 | XP_011520795.1 | 6 - 7 | 745 | 134 | |
XM_011522494.2 | XP_011520796.1 | 6 - 7 | 512 | 134 | |
GenBank mRNA | AF410154.1 | - | 6 - 7 | 624 | 134 |
AY084055.1 | - | 6 - 7 | 609 | 134 | |
AY699071.1 | - | 5 - 6 | 417 | 134 | |
U18259.1 | - | 6 - 7 | 624 | 134 | |
U31931.1 | - | 6 - 7 | 486 | 134 | |
X74301.1 | - | 6 - 7 | 601 | 134 |