The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016628.4 | NP_057712.2 | 6 - 7 | 696 | 72 |
NM_100264.2 | NP_567822.1 | 6 - 7 | 987 | 72 | |
XM_011519491.2 | XP_011517793.1 | 6 - 7 | 1297 | 72 | |
XM_017016315.1 | XP_016871804.1 | 6 - 7 | 807 | 72 | |
XM_017016316.1 | XP_016871805.1 | 6 - 7 | 1297 | 72 | |
GenBank mRNA | AB058747.1 | - | 4 - 5 | 406 | 72 |
AF116666.2 | - | 7 - 8 | 761 | 72 | |
AK055852.1 | - | 6 - 7 | 804 | 72 | |
AK223600.1 | - | 6 - 7 | 694 | 72 | |
AK225846.1 | - | 6 - 7 | 800 | 72 | |
AK290174.1 | - | 6 - 7 | 701 | 72 | |
AK297460.1 | - | 6 - 7 | 609 | 72 | |
AL713727.1 | - | 4 - 5 | 826 | 72 | |
BC004258.1 | - | 6 - 7 | 687 | 72 |