Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WAC
Entrez Gene ID
51322
Gene Name
WW domain containing adaptor with coiled-coil
Gene Aliases
BM-016, DESSH, PRO1741, Wwp4
Chromosome Location
Chr.10: 28532426 - 28623112 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016628.4 NP_057712.2 8 - 9 1258 85
NM_100264.2 NP_567822.1 8 - 9 1549 85
NM_100486.3 NP_567823.1 7 - 8 949 85
XM_011519491.2 XP_011517793.1 8 - 9 1859 85
XM_017016315.1 XP_016871804.1 8 - 9 1369 85
XM_017016316.1 XP_016871805.1 8 - 9 1859 85
XM_017016317.1 XP_016871806.1 7 - 8 1060 85
XM_017016318.1 XP_016871807.1 7 - 8 1060 85
GenBank mRNA AF208858.1 - 1 - 2 184 85
AK055852.1 - 8 - 9 1366 85
AK223600.1 - 8 - 9 1256 85
AK225846.1 - 8 - 9 1362 85
AK290174.1 - 8 - 9 1263 85
AK297460.1 - 9 - 10 1121 85
AL713727.1 - 6 - 7 1388 85
BC004258.1 - 8 - 9 1249 85
BC010356.1 - 3 - 4 400 85

View Full Product Details