The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105206.2 | NP_001098676.2 | 11 - 12 | 1757 | 55 |
NM_001105207.2 | NP_001098677.2 | 11 - 12 | 1753 | 55 | |
NM_002290.4 | NP_002281.3 | 11 - 12 | 1736 | 55 | |
XM_005266983.3 | XP_005267040.2 | 11 - 12 | 1630 | 55 | |
XM_005266984.3 | XP_005267041.2 | 11 - 12 | 1627 | 55 | |
XM_017010854.1 | XP_016866343.1 | 11 - 12 | 1606 | 55 | |
GenBank mRNA | AB210027.1 | - | 11 - 12 | 1609 | 55 |
AK027151.1 | - | 2 - 3 | 1552 | 55 | |
AK304401.1 | - | 11 - 12 | 1580 | 55 | |
BC066552.1 | - | 11 - 12 | 1749 | 55 | |
BX648467.1 | - | 6 - 7 | 791 | 55 | |
S78569.1 | - | 11 - 12 | 1528 | 55 | |
X76939.1 | - | 5 - 6 | 633 | 55 | |
X91171.1 | - | 11 - 12 | 1621 | 55 |