Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAMA4
Entrez Gene ID
3910
Gene Name
laminin subunit alpha 4
Gene Aliases
CMD1JJ, LAMA3, LAMA4*-1
Chromosome Location
Chr.6: 112107931 - 112254722 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105206.2 NP_001098676.2 15 - 16 2357 108
NM_001105207.2 NP_001098677.2 15 - 16 2353 108
NM_002290.4 NP_002281.3 15 - 16 2336 108
XM_005266983.3 XP_005267040.2 15 - 16 2230 108
XM_005266984.3 XP_005267041.2 15 - 16 2227 108
XM_017010854.1 XP_016866343.1 15 - 16 2206 108
GenBank mRNA AB210027.1 - 15 - 16 2209 108
AK304401.1 - 15 - 16 2180 108
BC066552.1 - 15 - 16 2349 108
BX648467.1 - 10 - 11 1391 108
S78569.1 - 15 - 16 2128 108
X76939.1 - 9 - 10 1233 108
X91171.1 - 15 - 16 2221 108

View Full Product Details