The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105206.2 | NP_001098676.2 | 28 - 29 | 4233 | 119 |
NM_001105207.2 | NP_001098677.2 | 28 - 29 | 4229 | 119 | |
NM_002290.4 | NP_002281.3 | 28 - 29 | 4212 | 119 | |
XM_005266983.3 | XP_005267040.2 | 28 - 29 | 4106 | 119 | |
XM_005266984.3 | XP_005267041.2 | 28 - 29 | 4103 | 119 | |
XM_017010854.1 | XP_016866343.1 | 28 - 29 | 4082 | 119 | |
GenBank mRNA | AB210027.1 | - | 28 - 29 | 4085 | 119 |
AK304401.1 | - | 28 - 29 | 4056 | 119 | |
BC066552.1 | - | 28 - 29 | 4225 | 119 | |
BX648467.1 | - | 23 - 24 | 3267 | 119 | |
S78569.1 | - | 28 - 29 | 4004 | 119 | |
X76939.1 | - | 22 - 23 | 3109 | 119 | |
X91171.1 | - | 28 - 29 | 4097 | 119 |