Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DNHD1
Entrez Gene ID
144132
Gene Name
dynein heavy chain domain 1
Gene Aliases
C11orf47, CCDC35, DHCD1, DNHD1L
Chromosome Location
Chr.11: 6497296 - 6572025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144666.2 NP_653267.2 15 - 16 3692 64
GenBank mRNA AB290171.1 - 2 - 3 380 64
AK074178.1 - 2 - 3 436 64

View Full Product Details