Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ORC5
- Entrez Gene ID
-
5001
- Gene Name
- origin recognition complex subunit 5
- Gene Aliases
-
ORC5L, ORC5P, ORC5T, PPP1R117
- Chromosome Location
-
Chr.7: 104126341 - 104208047 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK302122.1 |
- |
2 - 3 |
262 |
141 |
View Full Product Details