Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLAT
Entrez Gene ID
5327
Gene Name
plasminogen activator, tissue type
Gene Aliases
T-PA, TPA
Chromosome Location
Chr.8: 42174718 - 42207724 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000930.4 NP_000921.1 8 - 9 1063 84
NM_001319189.1 NP_001306118.1 6 - 7 796 84
NM_033011.3 NP_127509.1 7 - 8 925 84
GenBank mRNA A03776.1 - 7 - 8 840 84
A07197.1 - 7 - 8 830 84
AF260825.1 - 2 - 3 305 84
AK289387.1 - 8 - 9 1015 84
AK290575.1 - 8 - 9 1015 84
AK297738.1 - 6 - 7 637 84
AK297937.1 - 5 - 6 526 84
AK299205.1 - 8 - 9 905 84
AK300984.1 - 8 - 9 715 84
AK313342.1 - 7 - 8 766 84
AY221101.1 - 7 - 8 806 84
BC002795.2 - 7 - 8 751 84
BC007231.1 - 8 - 9 887 84
BC013968.2 - 8 - 9 889 84
BC018636.2 - 8 - 9 889 84
BT007060.1 - 7 - 8 806 84
D01096.1 - 6 - 7 718 84
FM956488.1 - 7 - 8 817 84
M15518.1 - 8 - 9 882 84
M18182.1 - 7 - 8 819 84
V00570.1 - 1 - 2 62 84
X02901.1 - 7 - 8 823 84
X07393.1 - 8 - 9 859 84

View Full Product Details