The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113361.1 | NP_001106832.1 | 10 - 11 | 1602 | 63 |
NM_001113363.1 | NP_001106834.1 | 8 - 9 | 868 | 63 | |
NM_001286805.1 | NP_001273734.1 | 8 - 9 | 737 | 63 | |
NM_020773.2 | NP_065824.2 | 10 - 11 | 1724 | 63 | |
XM_006713895.2 | XP_006713958.1 | 10 - 11 | 1892 | 63 | |
XM_011513507.2 | XP_011511809.1 | 11 - 12 | 1811 | 63 | |
XM_011513508.1 | XP_011511810.1 | 11 - 12 | 1704 | 63 | |
XM_011513513.2 | XP_011511815.1 | 9 - 10 | 906 | 63 | |
XM_017008478.1 | XP_016863967.1 | 9 - 10 | 974 | 63 | |
XM_017008480.1 | XP_016863969.1 | 11 - 12 | 1036 | 63 | |
GenBank mRNA | AB037743.1 | - | 9 - 10 | 1552 | 63 |
AB449900.1 | - | 9 - 10 | 1523 | 63 | |
AK295068.1 | - | 6 - 7 | 482 | 63 | |
AL833868.1 | - | 7 - 8 | 589 | 63 | |
BC041167.1 | - | 10 - 11 | 1647 | 63 |