Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PLAT
- Entrez Gene ID
-
5327
- Gene Name
- plasminogen activator, tissue type
- Gene Aliases
-
T-PA, TPA
- Chromosome Location
-
Chr.8: 42174718 - 42207724 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
X13097.1 |
- |
8 - 9 |
907 |
55 |
View Full Product Details