The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006948.4 | NP_008879.3 | 4 - 5 | 819 | 104 |
GenBank mRNA | AK128540.1 | - | 4 - 5 | 601 | 104 |
AK223282.1 | - | 4 - 5 | 777 | 104 | |
AK299004.1 | - | 3 - 4 | 554 | 104 | |
AK312396.1 | - | 4 - 5 | 774 | 104 | |
BC036370.2 | - | 4 - 5 | 781 | 104 | |
U04735.1 | - | 4 - 5 | 786 | 104 |