The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020156.4 | NP_064541.1 | 4 - 4 | 5254 | 114 |
XM_005249812.1 | XP_005249869.1 | 4 - 4 | 5322 | 114 | |
XM_011515453.2 | XP_011513755.1 | 5 - 5 | 5451 | 114 | |
XM_011515455.2 | XP_011513757.1 | 5 - 5 | 5373 | 114 | |
XM_011515456.2 | XP_011513758.1 | 4 - 4 | 5185 | 114 | |
XM_017012442.1 | XP_016867931.1 | 5 - 5 | 5373 | 114 | |
XM_017012443.1 | XP_016867932.1 | 5 - 5 | 5368 | 114 | |
XM_017012444.1 | XP_016867933.1 | 6 - 6 | 5359 | 114 | |
XM_017012445.1 | XP_016867934.1 | 7 - 7 | 5574 | 114 | |
XM_017012446.1 | XP_016867935.1 | 4 - 4 | 5203 | 114 | |
XM_017012447.1 | XP_016867936.1 | 5 - 5 | 5297 | 114 | |
XM_017012448.1 | XP_016867937.1 | 4 - 4 | 5208 | 114 | |
XM_017012449.1 | XP_016867938.1 | 3 - 3 | 5665 | 114 |