The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173560.3 | NP_775831.2 | 1 - 2 | 284 | 157 |
XM_011535589.1 | XP_011533891.1 | 1 - 2 | 279 | 157 | |
GenBank mRNA | BC039248.1 | - | 1 - 2 | 284 | 157 |
CA842450.1 | - | 1 - 2 | 215 | 157 |