Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEP44
Entrez Gene ID
80817
Gene Name
centrosomal protein 44
Gene Aliases
KIAA1712, PS1TP3
Chromosome Location
Chr.4: 174283677 - 174334355 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040157.2 NP_001035247.1 3 - 4 496 65
NM_001145314.1 NP_001138786.1 3 - 4 387 65
XM_011532285.2 XP_011530587.1 3 - 4 447 65
XM_011532286.2 XP_011530588.1 3 - 4 496 65
XM_011532287.2 XP_011530589.1 2 - 3 346 65
XM_017008654.1 XP_016864143.1 3 - 4 447 65
XM_017008655.1 XP_016864144.1 2 - 3 346 65
XM_017008656.1 XP_016864145.1 3 - 4 496 65
XM_017008657.1 XP_016864146.1 3 - 4 447 65
XM_017008658.1 XP_016864147.1 3 - 4 467 65
XM_017008659.1 XP_016864148.1 3 - 4 420 65
XM_017008660.1 XP_016864149.1 3 - 4 585 65
GenBank mRNA AB051499.1 - 3 - 4 253 65
AK094961.1 - 3 - 4 368 65
AK292484.1 - 3 - 4 368 65
AL699124.1 - 3 - 4 387 65
AY426674.1 - 1 - 2 82 65
BC038667.1 - 3 - 4 387 65
DB211405.1 - 3 - 4 468 65

View Full Product Details