The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005099.5 | NP_005090.3 | 8 - 9 | 2510 | 91 |
GenBank mRNA | AB014588.1 | - | 8 - 9 | 2484 | 91 |
AF148213.1 | - | 8 - 9 | 2490 | 91 | |
AK291573.1 | - | 8 - 9 | 2509 | 91 | |
AY358886.1 | - | 8 - 9 | 2502 | 91 | |
BC063293.1 | - | 8 - 9 | 2509 | 91 |