Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CALR
Entrez Gene ID
811
Gene Name
calreticulin
Gene Aliases
CRT, HEL-S-99n, RO, SSA, cC1qR
Chromosome Location
Chr.19: 12938600 - 12944490 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004343.3 NP_004334.1 1 - 2 177 100
GenBank mRNA AB451408.1 - 1 - 2 97 100
AK223060.1 - 1 - 2 65 100
AK295230.1 - - 120 100
AK304492.1 - - 124 100
AY047586.1 - 1 - 2 150 100
BC002500.2 - 1 - 2 130 100
BC007911.1 - 1 - 2 167 100
BC020493.1 - 1 - 2 159 100
BT007448.1 - 1 - 2 97 100
CR457070.1 - 1 - 2 97 100
FJ224311.1 - 1 - 2 163 100
M32294.1 - 1 - 2 163 100
M84739.1 - 1 - 2 205 100

View Full Product Details