Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD6
Entrez Gene ID
923
Gene Name
CD6 molecule
Gene Aliases
TP120
Chromosome Location
Chr.11: 60971641 - 61020377 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001254750.1 NP_001241679.1 4 - 5 1012 115
NM_001254751.1 NP_001241680.1 4 - 5 1012 115
NM_006725.4 NP_006716.3 4 - 5 1012 115
NR_045638.1 4 - 5 1012 115
XM_006718738.1 XP_006718801.1 4 - 5 1012 115
XM_006718739.1 XP_006718802.1 4 - 5 1012 115
XM_006718740.1 XP_006718803.1 4 - 5 1012 115
XM_006718741.1 XP_006718804.1 4 - 5 1012 115
XM_011545360.1 XP_011543662.1 4 - 5 1012 115
GenBank mRNA AK128419.1 - 5 - 6 916 115
AK292704.1 - 4 - 5 888 115
AK312749.1 - 4 - 5 973 115
BC033755.1 - 4 - 5 928 115
BC078669.1 - 4 - 5 951 115
U66142.1 - 4 - 5 787 115
U66145.1 - 4 - 5 787 115
U66146.1 - 4 - 5 787 115
X60992.1 - 4 - 5 907 115

View Full Product Details