Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMARCA4
Entrez Gene ID
6597
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 4
Gene Aliases
BAF190, BAF190A, BRG1, CSS4, MRD16, RTPS2, SNF2, SNF2L4, SNF2LB, SWI2, hSNF2b
Chromosome Location
Chr.19: 10960922 - 11062282 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128844.1 NP_001122316.1 6 - 7 1216 134
NM_001128845.1 NP_001122317.1 4 - 5 856 134
NM_001128846.1 NP_001122318.1 4 - 5 856 134
NM_001128847.1 NP_001122319.1 4 - 5 856 134
NM_001128848.1 NP_001122320.1 4 - 5 856 134
NM_001128849.1 NP_001122321.1 5 - 6 930 134
NM_003072.3 NP_003063.2 5 - 6 1140 134
XM_006722845.2 XP_006722908.1 5 - 6 1062 134
XM_006722846.2 XP_006722909.1 5 - 6 1156 134
XM_011528198.1 XP_011526500.1 6 - 7 1137 134
XM_017027160.1 XP_016882649.1 6 - 7 1137 134
XM_017027161.1 XP_016882650.1 6 - 7 1137 134
XM_017027162.1 XP_016882651.1 6 - 7 1137 134
XM_017027163.1 XP_016882652.1 6 - 7 1137 134
XM_017027164.1 XP_016882653.1 6 - 7 1137 134
XM_017027165.1 XP_016882654.1 6 - 7 1137 134
XM_017027166.1 XP_016882655.1 6 - 7 1137 134
XM_017027167.1 XP_016882656.1 6 - 7 1137 134
XM_017027168.1 XP_016882657.1 6 - 7 1137 134
GenBank mRNA AB209313.2 - 4 - 5 730 134
AK290033.1 - 6 - 7 1137 134
BC035916.1 - 5 - 6 990 134
BC136644.1 - 4 - 5 884 134
BC139917.1 - 5 - 6 1029 134
BC139924.1 - 5 - 6 1029 134
BC150298.1 - 5 - 6 1061 134
D26156.1 - 5 - 6 913 134
EU430756.1 - 4 - 5 856 134
EU430757.1 - 4 - 5 856 134
EU430758.1 - 4 - 5 856 134
EU430759.1 - 4 - 5 856 134
U29175.1 - 5 - 6 934 134

View Full Product Details