The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270941.1 | NP_001257870.1 | 11 - 12 | 2155 | 128 |
NM_001282282.1 | NP_001269211.1 | 10 - 11 | 1878 | 128 | |
NM_014790.4 | NP_055605.2 | 11 - 12 | 2155 | 128 | |
XM_017010097.1 | XP_016865586.1 | 12 - 13 | 2149 | 128 | |
XM_017010098.1 | XP_016865587.1 | 10 - 11 | 2035 | 128 | |
GenBank mRNA | AF273057.1 | - | 11 - 12 | 1996 | 128 |
AK291280.1 | - | 11 - 12 | 1877 | 128 | |
AK299724.1 | - | 10 - 11 | 1804 | 128 | |
BC017354.2 | - | 11 - 12 | 1920 | 128 | |
EF512550.1 | - | 10 - 11 | 1614 | 128 |