The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163334.1 | NP_001156806.1 | 16 - 17 | 2476 | 67 |
NM_001163335.1 | NP_001156807.1 | 17 - 18 | 2588 | 67 | |
NM_138780.2 | NP_620135.1 | 16 - 17 | 2497 | 67 | |
XM_011544001.2 | XP_011542303.1 | 19 - 20 | 2915 | 67 | |
XM_011544002.2 | XP_011542304.1 | 17 - 18 | 2549 | 67 | |
XM_017029972.1 | XP_016885461.1 | 20 - 21 | 3081 | 67 | |
XM_017029973.1 | XP_016885462.1 | 19 - 20 | 3148 | 67 | |
XM_017029974.1 | XP_016885463.1 | 18 - 19 | 2776 | 67 | |
GenBank mRNA | AB080222.1 | - | 15 - 16 | 2054 | 67 |
AK125106.1 | - | 16 - 17 | 2465 | 67 | |
BC131585.1 | - | 16 - 17 | 2120 | 67 | |
BX647688.1 | - | 17 - 18 | 2590 | 67 |