The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163334.1 | NP_001156806.1 | 1 - 2 | 473 | 100 |
NM_001163335.1 | NP_001156807.1 | 3 - 4 | 651 | 100 | |
NM_138780.2 | NP_620135.1 | 2 - 3 | 560 | 100 | |
XM_011544001.2 | XP_011542303.1 | 4 - 5 | 912 | 100 | |
XM_011544002.2 | XP_011542304.1 | 2 - 3 | 546 | 100 | |
XM_017029972.1 | XP_016885461.1 | 5 - 6 | 1078 | 100 | |
XM_017029973.1 | XP_016885462.1 | 4 - 5 | 1145 | 100 | |
XM_017029974.1 | XP_016885463.1 | 3 - 4 | 773 | 100 | |
GenBank mRNA | AB080222.1 | - | 1 - 2 | 117 | 100 |
AK125106.1 | - | 2 - 3 | 529 | 100 | |
BC131585.1 | - | 1 - 2 | 117 | 100 | |
BX647688.1 | - | 3 - 4 | 653 | 100 |