The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198581.2 | NP_940983.2 | 11 - 12 | 2477 | 86 |
XM_006712519.2 | XP_006712582.1 | 11 - 12 | 2484 | 86 | |
GenBank mRNA | AB111887.1 | - | 10 - 11 | 1923 | 86 |
AK123404.1 | - | 1 - 2 | 94 | 86 | |
AK127776.1 | - | 8 - 9 | 1546 | 86 | |
AK131416.1 | - | 11 - 12 | 2463 | 86 | |
BC155540.1 | - | 10 - 11 | 1923 | 86 | |
BC167154.1 | - | 10 - 11 | 1923 | 86 |