Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLBP
Entrez Gene ID
7884
Gene Name
stem-loop binding protein
Gene Aliases
HBP
Chromosome Location
Chr.4: 1692731 - 1712741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306074.1 NP_001293003.1 5 - 6 1077 73
NM_001306075.1 NP_001293004.1 5 - 6 1065 73
NM_006527.3 NP_006518.1 6 - 7 1182 73
GenBank mRNA AK091735.1 - 5 - 6 642 73
AK093314.1 - 6 - 7 655 73
AK094257.1 - 6 - 7 996 73
AK300826.1 - 5 - 6 655 73
AK315635.1 - 6 - 7 753 73
BC014908.1 - 6 - 7 751 73
BC015703.1 - 6 - 7 751 73
BT007433.1 - 6 - 7 629 73
U75679.1 - 6 - 7 744 73
Z71188.1 - 6 - 7 733 73

View Full Product Details