Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLBP
Entrez Gene ID
7884
Gene Name
stem-loop binding protein
Gene Aliases
HBP
Chromosome Location
Chr.4: 1692731 - 1712741 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306074.1 NP_001293003.1 6 - 7 1149 68
NM_001306075.1 NP_001293004.1 6 - 7 1137 68
NM_006527.3 NP_006518.1 7 - 8 1254 68
GenBank mRNA AK091735.1 - 6 - 7 714 68
AK093314.1 - 7 - 8 727 68
AK094257.1 - 7 - 8 1068 68
AK300826.1 - 6 - 7 727 68
AK315635.1 - 7 - 8 825 68
BC014908.1 - 7 - 8 823 68
BC015703.1 - 7 - 8 823 68
BT007433.1 - 7 - 8 701 68
U75679.1 - 7 - 8 816 68
Z71188.1 - 7 - 8 805 68

View Full Product Details