The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003320.4 | NP_003311.2 | 2 - 3 | 442 | 68 |
GenBank mRNA | BC075031.2 | - | 2 - 3 | 234 | 68 |
BC075032.2 | - | 2 - 3 | 234 | 68 | |
U82467.1 | - | 2 - 3 | 442 | 68 |