Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TUB
Entrez Gene ID
7275
Gene Name
tubby bipartite transcription factor
Gene Aliases
RDOB, rd5
Chromosome Location
Chr.11: 8019180 - 8106107 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003320.4 NP_003311.2 6 - 7 972 56
NM_177972.2 NP_813977.1 5 - 6 715 56
XM_005253109.3 XP_005253166.1 5 - 6 707 56
XM_011520344.2 XP_011518646.1 5 - 6 762 56
GenBank mRNA AK293528.1 - 5 - 6 643 56
BC075031.2 - 6 - 7 764 56
BC075032.2 - 6 - 7 764 56
U54644.1 - 5 - 6 718 56
U82467.1 - 6 - 7 972 56

View Full Product Details