Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CRNKL1
Entrez Gene ID
51340
Gene Name
crooked neck pre-mRNA splicing factor 1
Gene Aliases
CLF, CRN, Clf1, HCRN, MSTP021, SYF3
Chromosome Location
Chr.20: 20034361 - 20056046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278625.1 NP_001265554.1 14 - 15 2375 89
NM_001278626.1 NP_001265555.1 14 - 15 2188 89
NM_001278627.1 NP_001265556.1 14 - 15 2098 89
NM_001278628.1 NP_001265557.1 13 - 14 2046 89
NM_016652.5 NP_057736.4 14 - 15 2411 89
GenBank mRNA AB016840.1 - 13 - 14 2008 89
AF111802.1 - 13 - 14 2002 89
AF255443.2 - 14 - 15 2375 89
AF318302.1 - 13 - 14 1993 89
AF318303.1 - 14 - 15 2411 89
AF318304.1 - 14 - 15 2188 89
AF318305.1 - 14 - 15 2098 89
AK022908.1 - 12 - 13 1723 89
AK023246.1 - 10 - 11 1558 89
AK023728.1 - 9 - 10 1372 89
AK026342.1 - 3 - 4 432 89
AK292799.1 - 13 - 14 1916 89
AL832435.1 - 13 - 14 2020 89

View Full Product Details