The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001278625.1 | NP_001265554.1 | 9 - 10 | 1645 | 95 |
NM_001278626.1 | NP_001265555.1 | 9 - 10 | 1458 | 95 | |
NM_001278627.1 | NP_001265556.1 | 9 - 10 | 1368 | 95 | |
NM_001278628.1 | NP_001265557.1 | 8 - 9 | 1316 | 95 | |
NM_016652.5 | NP_057736.4 | 9 - 10 | 1681 | 95 | |
GenBank mRNA | AB016840.1 | - | 8 - 9 | 1278 | 95 |
AF111802.1 | - | 8 - 9 | 1272 | 95 | |
AF255443.2 | - | 9 - 10 | 1645 | 95 | |
AF318302.1 | - | 8 - 9 | 1263 | 95 | |
AF318303.1 | - | 9 - 10 | 1681 | 95 | |
AF318304.1 | - | 9 - 10 | 1458 | 95 | |
AF318305.1 | - | 9 - 10 | 1368 | 95 | |
AK022908.1 | - | 7 - 8 | 993 | 95 | |
AK023246.1 | - | 5 - 6 | 828 | 95 | |
AK023728.1 | - | 4 - 5 | 642 | 95 | |
AK292799.1 | - | 8 - 9 | 1186 | 95 | |
AL832435.1 | - | 8 - 9 | 1290 | 95 |