The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004747.3 | NP_004738.3 | 21 - 22 | 4259 | 98 |
XM_005270276.4 | XP_005270333.1 | 21 - 22 | 4213 | 98 | |
XM_006718056.3 | XP_006718119.1 | 20 - 21 | 3202 | 98 | |
XM_011540341.2 | XP_011538643.1 | 21 - 22 | 4383 | 98 | |
XM_011540342.1 | XP_011538644.1 | 21 - 22 | 3961 | 98 | |
XM_011540344.2 | XP_011538646.1 | 20 - 21 | 3868 | 98 | |
XM_011540345.1 | XP_011538647.1 | 20 - 21 | 3993 | 98 | |
XM_011540346.2 | XP_011538648.1 | 21 - 22 | 4228 | 98 | |
XM_011540347.2 | XP_011538649.1 | 17 - 18 | 3439 | 98 | |
XM_017016913.1 | XP_016872402.1 | 21 - 22 | 3922 | 98 | |
XM_017016914.1 | XP_016872403.1 | 21 - 22 | 4252 | 98 | |
XM_017016915.1 | XP_016872404.1 | 17 - 18 | 4123 | 98 | |
GenBank mRNA | AB011155.2 | - | 21 - 22 | 4289 | 98 |
AB091676.1 | - | 19 - 20 | 3724 | 98 | |
AK122931.1 | - | 22 - 23 | 4191 | 98 | |
AK314771.1 | - | 5 - 6 | 568 | 98 | |
BC073996.1 | - | 7 - 8 | 969 | 98 | |
BC117695.2 | - | 11 - 12 | 4017 | 98 | |
BC146794.1 | - | 21 - 22 | 4289 | 98 | |
BX649131.1 | - | 16 - 17 | 2366 | 98 | |
U61843.1 | - | 6 - 7 | 817 | 98 |