The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032312.3 | NP_115688.1 | 2 - 3 | 503 | 116 |
XM_005264599.3 | XP_005264656.1 | 2 - 3 | 497 | 116 | |
XM_011533125.2 | XP_011531427.1 | 2 - 3 | 497 | 116 | |
GenBank mRNA | AK098486.1 | - | 2 - 3 | 482 | 116 |
BC004875.2 | - | 2 - 3 | 430 | 116 |