The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133170.3 | NP_573400.3 | 14 - 15 | 2422 | 105 |
XM_017027612.1 | XP_016883101.1 | 14 - 15 | 2485 | 105 | |
GenBank mRNA | AF043644.4 | - | 14 - 15 | 2422 | 105 |