Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTPRT
Entrez Gene ID
11122
Gene Name
protein tyrosine phosphatase, receptor type T
Gene Aliases
RPTPrho
Chromosome Location
Chr.20: 42072752 - 43189917 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007050.5 NP_008981.4 26 - 27 3898 79
NM_133170.3 NP_573400.3 27 - 28 3955 79
XM_017027611.1 XP_016883100.1 26 - 27 3991 79
XM_017027612.1 XP_016883101.1 26 - 27 3988 79
XM_017027613.1 XP_016883102.1 25 - 26 3930 79
GenBank mRNA AB006621.2 - 26 - 27 3806 79
AF043644.4 - 27 - 28 3964 79
BC153300.1 - 26 - 27 3806 79

View Full Product Details