Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR2D
Entrez Gene ID
5433
Gene Name
RNA polymerase II subunit D
Gene Aliases
HSRBP4, HSRPB4, RBP4, RPB16
Chromosome Location
Chr.2: 127846266 - 127858155 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004805.3 NP_004796.1 1 - 2 133 93
GenBank mRNA BC017205.2 - 1 - 2 105 93
BC093795.1 - 1 - 2 133 93
BC093797.1 - 1 - 2 135 93
U85510.1 - 1 - 2 101 93

View Full Product Details