The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004805.3 | NP_004796.1 | 3 - 4 | 403 | 82 |
GenBank mRNA | BC017205.2 | - | 3 - 4 | 375 | 82 |
BC093795.1 | - | 3 - 4 | 403 | 82 | |
BC093797.1 | - | 3 - 4 | 405 | 82 | |
U85510.1 | - | 3 - 4 | 371 | 82 |