The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005073.3 | NP_005064.1 | 6 - 7 | 539 | 113 |
GenBank mRNA | AB001328.1 | - | 4 - 5 | 558 | 113 |
AF043233.1 | - | 6 - 7 | 518 | 113 | |
AY634368.1 | - | 6 - 7 | 518 | 113 | |
BC096327.1 | - | 6 - 7 | 493 | 113 | |
BC096328.3 | - | 6 - 7 | 493 | 113 | |
BC096329.3 | - | 6 - 7 | 493 | 113 | |
EU544224.1 | - | 5 - 6 | 462 | 113 | |
U13173.1 | - | 6 - 7 | 518 | 113 | |
U21936.1 | - | 6 - 7 | 518 | 113 |