Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH3
Entrez Gene ID
55567
Gene Name
dynein axonemal heavy chain 3
Gene Aliases
DLP3, DNAHC3B, HEL-36, Hsadhc3, dnahc3-b
Chromosome Location
Chr.16: 20933111 - 21159537 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017539.2 NP_060009.1 3 - 4 446 139
XM_006721062.1 XP_006721125.1 4 - 5 635 139
XM_011545882.2 XP_011544184.1 5 - 6 1075 139
XM_011545883.1 XP_011544185.1 5 - 6 843 139
XM_011545885.2 XP_011544187.1 5 - 6 1075 139
XM_017023426.1 XP_016878915.1 5 - 6 1075 139
XM_017023427.1 XP_016878916.1 5 - 6 1075 139
XM_017023428.1 XP_016878917.1 5 - 6 1075 139
XM_017023429.1 XP_016878918.1 5 - 6 1076 139
XM_017023431.1 XP_016878920.1 5 - 6 1076 139
XM_017023432.1 XP_016878921.1 3 - 4 542 139
GenBank mRNA AF494040.1 - 3 - 4 446 139
AK056509.2 - 4 - 5 635 139
AK303958.1 - 4 - 5 644 139
AK310973.1 - 4 - 5 632 139

View Full Product Details