Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ABCB9
Entrez Gene ID
23457
Gene Name
ATP binding cassette subfamily B member 9
Gene Aliases
EST122234, TAPL
Chromosome Location
Chr.12: 122918708 - 122975160 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243013.1 NP_001229942.1 1 - 2 226 114
NM_001243014.1 NP_001229943.1 1 - 2 226 114
NM_019624.3 NP_062570.1 1 - 2 226 114
NM_019625.3 NP_062571.1 1 - 2 226 114
NM_203444.3 NP_982269.2 1 - 2 226 114
XM_005253558.2 XP_005253615.1 1 - 2 180 114
GenBank mRNA AB112582.1 - 1 - 2 177 114
AB112583.1 - 1 - 2 177 114
AB177852.1 - 1 - 2 138 114
AF216494.1 - 1 - 2 214 114
AK027624.1 - 1 - 2 119 114
AK304295.1 - 1 - 2 222 114
DA796102.1 - 1 - 2 226 114

View Full Product Details