Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PYGL
Entrez Gene ID
5836
Gene Name
phosphorylase, glycogen, liver
Gene Aliases
GSD6
Chromosome Location
Chr.14: 50905217 - 50944530 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163940.1 NP_001157412.1 7 - 8 1022 74
NM_002863.4 NP_002854.3 8 - 9 1124 74
GenBank mRNA AF046785.1 - 8 - 9 997 74
AF066858.1 - 8 - 9 997 74
AK225904.1 - 8 - 9 1077 74
AK300580.1 - 7 - 8 975 74
AK309333.1 - 8 - 9 1050 74
AK313206.1 - 8 - 9 1038 74
AY429567.1 - 4 - 5 383 74
BC009895.2 - 8 - 9 1067 74
BC082229.1 - 8 - 9 1068 74
BC095850.1 - 8 - 9 1084 74
BC110791.1 - 8 - 9 1066 74
Y15233.1 - 8 - 9 1048 74

View Full Product Details