Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRPX
Entrez Gene ID
8406
Gene Name
sushi repeat containing protein, X-linked
Gene Aliases
DRS, ETX1, HEL-S-83p, SRPX1
Chromosome Location
Chr.X: 38149335 - 38220924 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170750.1 NP_001164221.1 3 - 4 595 83
NM_001170752.1 NP_001164223.1 4 - 5 655 83
NM_006307.4 NP_006298.1 4 - 5 655 83
XM_017029893.1 XP_016885382.1 4 - 5 594 83
GenBank mRNA AK125542.1 - 3 - 4 576 83
AK289430.1 - 4 - 5 602 83
AK293127.1 - 4 - 5 632 83
BC020684.1 - 4 - 5 610 83
U78093.1 - 4 - 5 562 83

View Full Product Details