The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014316.3 | NP_055131.2 | 1 - 2 | 44 | 93 |
GenBank mRNA | AK093698.1 | - | 1 - 2 | 38 | 93 |
AK311777.1 | - | 1 - 2 | 38 | 93 | |
BC003366.1 | - | 1 - 2 | 41 | 93 | |
BC108283.1 | - | 1 - 2 | 24 | 93 | |
HY090855.1 | - | 1 - 2 | 44 | 93 |