The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012308.2 | NP_036440.1 | 3 - 4 | 1037 | 91 |
NR_027473.1 | 3 - 4 | 1037 | 91 | ||
XM_011544860.2 | XP_011543162.1 | 3 - 4 | 1096 | 91 | |
XM_017017391.1 | XP_016872880.1 | 3 - 4 | 343 | 91 | |
XM_017017392.1 | XP_016872881.1 | 2 - 3 | 154 | 91 | |
GenBank mRNA | AB023221.2 | - | 3 - 4 | 354 | 91 |
AK024505.1 | - | 3 - 4 | 279 | 91 | |
BC047486.1 | - | 3 - 4 | 1039 | 91 | |
BC064360.1 | - | 3 - 4 | 452 | 91 | |
JQ710741.1 | - | 2 - 3 | 221 | 91 | |
JQ710742.1 | - | 3 - 4 | 343 | 91 | |
JQ710743.1 | - | 2 - 3 | 221 | 91 |