The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319053.1 | NP_001305982.1 | 8 - 9 | 1192 | 68 |
NM_001470.3 | NP_001461.1 | 8 - 9 | 1256 | 68 | |
NM_021903.2 | NP_068703.1 | 3 - 4 | 1056 | 68 | |
NM_021904.3 | NP_068704.2 | 7 - 8 | 1070 | 68 | |
XM_005248982.2 | XP_005249039.1 | 8 - 9 | 1071 | 68 | |
XM_006715047.3 | XP_006715110.1 | 7 - 8 | 871 | 68 | |
XM_011514453.2 | XP_011512755.1 | 6 - 7 | 850 | 68 | |
XM_011514455.1 | XP_011512757.1 | 8 - 9 | 1071 | 68 | |
XM_017010676.1 | XP_016866165.1 | 4 - 5 | 602 | 68 | |
GenBank mRNA | AB208790.1 | - | 8 - 9 | 991 | 68 |
AF099148.1 | - | 8 - 9 | 1078 | 68 | |
AF301005.1 | - | 7 - 8 | 973 | 68 | |
AJ012185.1 | - | 7 - 8 | 970 | 68 | |
AJ012186.1 | - | 3 - 4 | 619 | 68 | |
AJ012187.1 | - | 6 - 7 | 784 | 68 | |
AJ012288.1 | - | 8 - 9 | 1013 | 68 | |
AJ225028.1 | - | 8 - 9 | 1204 | 68 | |
AJ225029.1 | - | 3 - 4 | 798 | 68 | |
AK223619.1 | - | 3 - 4 | 841 | 68 | |
AK290191.1 | - | 3 - 4 | 841 | 68 | |
AK312308.1 | - | 3 - 4 | 837 | 68 | |
BC041332.2 | - | 3 - 4 | 841 | 68 | |
BC042598.1 | - | 3 - 4 | 830 | 68 | |
BC050532.1 | - | 3 - 4 | 828 | 68 | |
Y11044.1 | - | 7 - 8 | 967 | 68 |