Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NR1D2
Entrez Gene ID
9975
Gene Name
nuclear receptor subfamily 1 group D member 2
Gene Aliases
BD73, EAR-1R, RVR
Chromosome Location
Chr.3: 23945260 - 23980618 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145425.1 NP_001138897.1 4 - 5 633 66
NM_005126.4 NP_005117.3 4 - 5 835 66
NR_110524.1 4 - 5 835 66
XM_006713451.3 XP_006713514.1 4 - 5 570 66
GenBank mRNA AB209091.1 - 3 - 4 423 66
AB307693.1 - 4 - 5 516 66
AK301925.1 - 4 - 5 633 66
AK313464.1 - 5 - 6 653 66
BC045613.1 - 4 - 5 783 66
BC070035.1 - 4 - 5 722 66
CR457434.1 - 4 - 5 516 66
D16815.1 - 4 - 5 821 66
HQ692830.1 - 4 - 5 516 66
HQ692831.1 - 3 - 4 291 66

View Full Product Details