Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBE3A
Entrez Gene ID
7337
Gene Name
ubiquitin protein ligase E3A
Gene Aliases
ANCR, AS, E6-AP, EPVE6AP, HPVE6A
Chromosome Location
Chr.15: 25337234 - 25439086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000462.3 NP_000453.2 7 - 8 2374 78
NM_130838.1 NP_570853.1 3 - 4 1589 78
NM_130839.2 NP_570854.1 6 - 7 2309 78
XM_005268267.4 XP_005268324.1 8 - 9 2676 78
XM_005268268.4 XP_005268325.1 7 - 8 2563 78
XM_005268269.4 XP_005268326.1 8 - 9 2407 78
XM_005268270.4 XP_005268327.1 7 - 8 2364 78
XM_005268271.4 XP_005268328.1 4 - 5 2183 78
XM_006720675.3 XP_006720738.1 8 - 9 2628 78
XM_006720676.3 XP_006720739.1 5 - 6 1873 78
XM_011521994.2 XP_011520296.1 8 - 9 2565 78
XM_011521995.2 XP_011520297.1 8 - 9 2239 78
XM_017022544.1 XP_016878033.1 9 - 10 2601 78
XM_017022545.1 XP_016878034.1 8 - 9 2560 78
XM_017022546.1 XP_016878035.1 8 - 9 2468 78
XM_017022547.1 XP_016878036.1 7 - 8 2500 78
XM_017022548.1 XP_016878037.1 7 - 8 2495 78
XM_017022549.1 XP_016878038.1 7 - 8 2302 78
XM_017022550.1 XP_016878039.1 7 - 8 2454 78
XM_017022551.1 XP_016878040.1 8 - 9 2505 78
XM_017022552.1 XP_016878041.1 8 - 9 2195 78
XM_017022553.1 XP_016878042.1 8 - 9 2510 78
XM_017022554.1 XP_016878043.1 4 - 5 2514 78
XM_017022555.1 XP_016878044.1 6 - 7 2283 78
XM_017022556.1 XP_016878045.1 7 - 8 2432 78
XM_017022557.1 XP_016878046.1 6 - 7 2367 78
XM_017022558.1 XP_016878047.1 7 - 8 2563 78
XM_017022559.1 XP_016878048.1 4 - 5 2183 78
GenBank mRNA AB194232.1 - 5 - 6 1685 78
AF273050.1 - 4 - 5 1644 78
AK130491.1 - 2 - 3 1318 78
AK291405.1 - 4 - 5 1735 78
AK292514.1 - 6 - 7 1919 78
BC002582.2 - 6 - 7 2262 78
BC009271.1 - 1 - 2 866 78
L07557.1 - 3 - 4 1611 78
U84404.1 - 8 - 9 2131 78
X98031.1 - 4 - 5 1614 78
X98032.1 - 2 - 3 1545 78
X98033.1 - 4 - 5 1605 78

View Full Product Details