The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139488.1 | NP_001132960.1 | 15 - 16 | 2288 | 88 |
NM_015376.2 | NP_056191.1 | 14 - 15 | 1843 | 88 | |
NM_170672.2 | NP_733772.1 | 16 - 17 | 2201 | 88 | |
XM_005264246.3 | XP_005264303.1 | 15 - 16 | 2625 | 88 | |
XM_011532746.2 | XP_011531048.1 | 16 - 17 | 2075 | 88 | |
XM_011532747.2 | XP_011531049.1 | 15 - 16 | 2090 | 88 | |
XM_011532748.2 | XP_011531050.2 | 15 - 16 | 1936 | 88 | |
XM_017003759.1 | XP_016859248.1 | 17 - 18 | 3551 | 88 | |
XM_017003760.1 | XP_016859249.1 | 16 - 17 | 4023 | 88 | |
XM_017003761.1 | XP_016859250.1 | 15 - 16 | 1953 | 88 | |
XM_017003762.1 | XP_016859251.1 | 16 - 17 | 2199 | 88 | |
XM_017003763.1 | XP_016859252.1 | 15 - 16 | 1929 | 88 | |
GenBank mRNA | AB020653.1 | - | 14 - 15 | 1843 | 88 |
AK292983.1 | - | 16 - 17 | 2112 | 88 | |
BC027849.1 | - | 16 - 17 | 2201 | 88 | |
BX647990.1 | - | 15 - 16 | 1896 | 88 |