The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286572.2 | NP_001273501.1 | 1 - 2 | 111 | 67 |
GenBank mRNA | AK297531.1 | - | 1 - 2 | 80 | 67 |
CR627431.1 | - | 1 - 2 | 79 | 67 | |
DA328615.1 | - | 1 - 2 | 111 | 67 |